Gene Transcripts

Gene Symbol
ADAM5
Entrez Gene ID
255926
Gene Name
ADAM metallopeptidase domain 5 (pseudogene)
Gene Aliases
ADAM5P, TMDCII
Chromosome Location
Chr. 8: 39314663 - 39417226 on Build GRCh38
UniGene ID
Hs.369765
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001448.1 9 784
GenBank mRNA AJ132821.1 - 8 718
AJ132822.1 - 8 600
AJ132823.1 - 7 534
BC047448.1 - 9 918
BC067864.1 - 7 577

View Full Product Details