Gene Transcripts
- Gene Symbol
- CDCA5
- Entrez Gene ID
- 113130
- Gene Name
- cell division cycle associated 5
- Gene Aliases
- SORORIN
- Chromosome Location
- Chr. 11: 65061009 - 65084143 on Build GRCh38
- UniGene ID
- Hs.434886
- Species
- Homo sapiens
- Gene Symbol
- LOC256676
- Entrez Gene ID
- 256676
- Gene Name
- uncharacterized LOC256676
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.733372
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB231703.1 |
- |
2 |
93 |
|
View Full Product Details