Gene Transcripts
- Gene Symbol
- C19orf54
- Entrez Gene ID
- 284325
- Gene Name
- chromosome 19 open reading frame 54
- Gene Aliases
- Chromosome Location
- Chr. 19: 40740856 - 40750713 on Build GRCh38
- UniGene ID
- Hs.585105
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_198476.3 |
NP_940878.3
|
4, 5 |
776 |
|
XM_005258776.4 |
XP_005258833.1
|
4, 5 |
1356 |
|
XM_006723152.2 |
XP_006723215.1
|
4, 5 |
1356 |
|
XM_006723153.2 |
XP_006723216.1
|
4, 5 |
1356 |
|
XM_011526774.2 |
XP_011525076.1
|
4, 5 |
1356 |
|
XM_011526775.1 |
XP_011525077.1
|
4, 5 |
1356 |
|
XM_011526776.2 |
XP_011525078.1
|
4, 5 |
1356 |
|
XM_011526777.2 |
XP_011525079.1
|
4, 5 |
1356 |
|
XM_011526778.2 |
XP_011525080.1
|
4, 5 |
1356 |
|
XM_011526779.2 |
XP_011525081.1
|
4, 5 |
1356 |
|
XM_011526782.2 |
XP_011525084.1
|
4, 5 |
1356 |
|
XM_011526784.2 |
XP_011525086.1
|
5, 6 |
679 |
|
XM_011526786.2 |
XP_011525088.1
|
4, 5 |
1356 |
|
XM_017026620.1 |
XP_016882109.1
|
4, 5 |
1356 |
|
XM_017026621.1 |
XP_016882110.1
|
4, 5 |
1356 |
|
XM_017026622.1 |
XP_016882111.1
|
4, 5 |
1356 |
|
XM_017026624.1 |
XP_016882113.1
|
5, 6 |
679 |
|
XM_017026625.1 |
XP_016882114.1
|
4, 5 |
1356 |
|
XM_017026626.1 |
XP_016882115.1
|
5, 6 |
679 |
|
GenBank mRNA |
AK123126.1 |
- |
5, 6 |
679 |
|
BI521309.1 |
- |
5, 6 |
608 |
|
CN281611.1 |
- |
4, 5 |
439 |
|
View Full Product Details