Gene Transcripts
- Gene Symbol
- WHAMMP3
- Entrez Gene ID
- 339005
- Gene Name
- WAS protein homolog associated with actin, golgi membranes and microtubules pseudogene 3
- Gene Aliases
- WHAMML1, WHDC1L1
- Chromosome Location
- Chr. 15: 22664739 - 22685368 on Build GRCh38
- UniGene ID
- Hs.212670
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003521.1 |
|
11 |
3453 |
|
GenBank mRNA |
BC048987.1 |
- |
11 |
3514 |
|
DQ309040.1 |
- |
11 |
3453 |
|
- Gene Symbol
- WHAMMP2
- Entrez Gene ID
- 440253
- Gene Name
- WAS protein homolog associated with actin, golgi membranes and microtubules pseudogene 2
- Gene Aliases
- WHAMML2, WHDC1L2
- Chromosome Location
- Chr. 15: 28737583 - 28758362 on Build GRCh38
- UniGene ID
- Hs.558967
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026589.1 |
|
9 |
5108 |
|
View Full Product Details