Gene Transcripts

Gene Symbol
H1FNT
Entrez Gene ID
341567
Gene Name
H1 histone family member N, testis specific
Gene Aliases
H1.7, H1T2
Chromosome Location
Chr. 12: 48328980 - 48330279 on Build GRCh38
UniGene ID
Hs.155833
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181788.1 NP_861453.1 1 329
GenBank mRNA AY302593.1 - 1 329
BC118635.1 - 1 277 40023772
BC119787.1 - 1 279 40023773

View Full Product Details