Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001129820.1 | NP_001123292.1 | 1 | 707 | |
XM_017024576.1 | XP_016880065.1 | 3 | 876 | ||
XM_017024577.1 | XP_016880066.1 | 3 | 898 | ||
XM_017024578.1 | XP_016880067.1 | 2 | 765 | ||
XM_017024579.1 | XP_016880068.1 | 2 | 798 | ||
GenBank mRNA | BC140847.1 | - | 1 | 704 | |
BC157877.1 | - | 1 | 708 | 9057094 | |
BC157879.1 | - | 1 | 715 | 9057095 |