Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001195545.1 | NP_001182474.1 | 2 | 210 | |
XM_011524158.2 | XP_011522460.1 | 4 | 396 | ||
XM_017024003.1 | XP_016879492.1 | 4 | 375 | ||
GenBank mRNA | DY655553.1 | - | 2 | 210 |