Gene Transcripts
- Gene Symbol
- PI4KAP2
- Entrez Gene ID
- 375133
- Gene Name
- phosphatidylinositol 4-kinase alpha pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 22: 21472998 - 21517491 on Build GRCh38
- UniGene ID
- Hs.448225
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC139841.1 |
- |
11, 12 |
1280 |
|
CR456378.1 |
- |
11, 12 |
1298 |
|
- Gene Symbol
- PI4KAP1
- Entrez Gene ID
- 728233
- Gene Name
- phosphatidylinositol 4-kinase alpha pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 22: 18533864 - 18548828 on Build GRCh38
- UniGene ID
- Hs.592456
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003563.1 |
|
11, 12 |
1232 |
|
GenBank mRNA |
AK292695.1 |
- |
12, 13 |
1323 |
|
BC131538.1 |
- |
11, 12 |
1232 |
|
View Full Product Details