Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_198694.3 | NP_941967.3 | 1 | 870 | |
GenBank mRNA | AB076352.1 | - | 1 | 870 | |
AJ566384.1 | - | 1 | 66 | ||
BC120949.1 | - | 1 | 869 | 40115143 | |
BC120950.1 | - | 1 | 869 | 40115146 |