Gene Transcripts

Gene Symbol
GVINP1
Entrez Gene ID
387751
Gene Name
GTPase, very large interferon inducible pseudogene 1
Gene Aliases
GVIN1, GVIN1P, VLIG-1, VLIG1
Chromosome Location
Chr. 11: 6713139 - 6721879 on Build GRCh38
UniGene ID
Hs.494757
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003945.1 1 7047
GenBank mRNA AK023435.1 - 1 1632
AL834507.1 - 1 1698
BX538342.1 - 1 3149

View Full Product Details