Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278556.1 | NP_001265485.1 | 5 | 426 | |
NM_001287222.1 | NP_001274151.1 | 5 | 426 | ||
GenBank mRNA | AF004561.1 | - | 4 | 275 | |
AF006086.1 | - | 5 | 300 | ||
AK297309.1 | - | 5 | 357 | ||
AK311789.1 | - | 5 | 373 | ||
AL357196.1 | - | 5 | 336 | ||
BC067747.1 | - | 5 | 332 | 5302338 | |
BC078162.1 | - | 5 | 309 | 6148231 | |
BG620669.1 | - | 5 | 344 | ||
BG723870.1 | - | 5 | 390 | ||
BM999380.1 | - | 3 | 530 | ||
CR407667.1 | - | 4 | 275 | ||
DQ328220.1 | - | 5 | 356 |