Gene Transcripts

Gene Symbol
SLC22A31
Entrez Gene ID
146429
Gene Name
solute carrier family 22 member 31
Gene Aliases
Chromosome Location
Chr. 16: 89195756 - 89201674 on Build GRCh38
UniGene ID
Hs.447544
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242757.1 NP_001229686.1 3 705
XM_011522897.2 XP_011521199.1 4 753
XM_011522899.2 XP_011521201.1 4 690
XM_017022962.1 XP_016878451.1 1 355
XM_017022963.1 XP_016878452.1 2 1878
XM_017022964.1 XP_016878453.1 3 1034
XM_017022965.1 XP_016878454.1 2 1819
XM_017022966.1 XP_016878455.1 3 1657

View Full Product Details