Gene Transcripts

Gene Symbol
KRTDAP
Entrez Gene ID
388533
Gene Name
keratinocyte differentiation associated protein
Gene Aliases
KDAP, UNQ467
Chromosome Location
Chr. 19: 35487324 - 35490531 on Build GRCh38
UniGene ID
Hs.112457
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207392.2 NP_997275.1 3, 4 251
GenBank mRNA AJ293408.1 - 3, 4 212
AY359007.1 - 3, 4 174
BC130501.1 - 3, 4 251 40146532

View Full Product Details