Gene Transcripts

Gene Symbol
CCDC61
Entrez Gene ID
729440
Gene Name
coiled-coil domain containing 61
Gene Aliases
Chromosome Location
Chr. 19: 45995457 - 46018616 on Build GRCh38
UniGene ID
Hs.515479
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267723.1 NP_001254652.1 7 877
XM_005259191.3 XP_005259248.1 7 2982
XM_005259192.4 XP_005259249.1 7 1623
XM_011527257.2 XP_011525559.1 7 906
GenBank mRNA AW453013.1 - 2 140
BI819372.1 - 6 780

View Full Product Details