Gene Transcripts
- Gene Symbol
- CENPN
- Entrez Gene ID
- 55839
- Gene Name
- centromere protein N
- Gene Aliases
- BM039, C16orf60, CENP-N, ICEN32
- Chromosome Location
- Chr. 16: 81006498 - 81033107 on Build GRCh38
- UniGene ID
- Hs.726537
- Species
- Homo sapiens
- Gene Symbol
- GATS
- Entrez Gene ID
- 352954
- Gene Name
- GATS, stromal antigen 3 opposite strand
- Gene Aliases
- STAG3OS
- Chromosome Location
- Chr. 7: 100187988 - 100272274 on Build GRCh38
- UniGene ID
- Hs.556063
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124689.1 |
- |
2 |
3757 |
|
- Gene Symbol
- HEATR3
- Entrez Gene ID
- 55027
- Gene Name
- HEAT repeat containing 3
- Gene Aliases
- SYO1
- Chromosome Location
- Chr. 16: 50065970 - 50107268 on Build GRCh38
- UniGene ID
- Hs.733027
- Species
- Homo sapiens
- Gene Symbol
- LOC105371242
- Entrez Gene ID
- 105371242
- Gene Name
- peptidyl-prolyl cis-trans isomerase A-like 4C
- Gene Aliases
- Chromosome Location
- Chr. 1: 146342335 - 146344907 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- PPIAL4C
- Entrez Gene ID
- 653598
- Gene Name
- peptidylprolyl isomerase A like 4C
- Gene Aliases
- COAS-2
- Chromosome Location
- Chr. 1: 149583823 - 149584607 on Build GRCh38
- UniGene ID
- Hs.534674
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK123006.1 |
- |
2 |
2753 |
|
- Gene Symbol
- SLX1A-SULT1A3
- Entrez Gene ID
- 100526830
- Gene Name
- SLX1A-SULT1A3 readthrough (NMD candidate)
- Gene Aliases
- Chromosome Location
- Chr. 16: 30194433 - 30204329 on Build GRCh38
- UniGene ID
- Hs.460558
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK094769.1 |
- |
1 |
252 |
|
View Full Product Details