Gene Transcripts

Gene Symbol
PRR23B
Entrez Gene ID
389151
Gene Name
proline rich 23B
Gene Aliases
Chromosome Location
Chr. 3: 139019031 - 139020926 on Build GRCh38
UniGene ID
Hs.531376
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013650.2 NP_001013672.1 1 672
GenBank mRNA AK127998.1 - 1 672
BC137145.1 - 1 660 9021142
BC137146.1 - 1 658 9021143

View Full Product Details