Gene Transcripts

Gene Symbol
PRR23C
Entrez Gene ID
389152
Gene Name
proline rich 23C
Gene Aliases
Chromosome Location
Chr. 3: 139042102 - 139044892 on Build GRCh38
UniGene ID
Hs.531377
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134657.1 NP_001128129.1 1 923
GenBank mRNA AK128089.1 - 1 920

View Full Product Details