Gene Transcripts

Gene Symbol
C5orf52
Entrez Gene ID
100190949
Gene Name
chromosome 5 open reading frame 52
Gene Aliases
Chromosome Location
Chr. 5: 157671237 - 157680176 on Build GRCh38
UniGene ID
Hs.720862
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145132.1 NP_001138604.1 1, 2 264
XM_011534416.1 XP_011532718.1 2, 3 290
GenBank mRNA BG721329.1 - 1, 2 87

View Full Product Details