Gene Transcripts
- Gene Symbol
- IKZF1
- Entrez Gene ID
- 10320
- Gene Name
- IKAROS family zinc finger 1
- Gene Aliases
- CVID13, Hs.54452, IK1, IKAROS, LYF1, LyF-1, PPP1R92, PRO0758, ZNFN1A1
- Chromosome Location
- Chr. 7: 50303453 - 50405101 on Build GRCh38
- UniGene ID
- Hs.435949
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF116605.1 |
- |
1 |
830 |
|
View Full Product Details