Gene Transcripts

Gene Symbol
DNAJB3
Entrez Gene ID
414061
Gene Name
DnaJ heat shock protein family (Hsp40) member B3
Gene Aliases
HCG3
Chromosome Location
Chr. 2: 233742750 - 233744015 on Build GRCh38
UniGene ID
Hs.725533
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001394.3 NP_001001394.1 1 280
GenBank mRNA AY083665.1 - 1 280
BC017590.2 - 1 280 4828023
BC024013.2 - 1 243 4826480

View Full Product Details