Gene Transcripts
- Gene Symbol
- FAM96A
- Entrez Gene ID
- 84191
- Gene Name
- family with sequence similarity 96 member A
- Gene Aliases
- Chromosome Location
- Chr. 15: 64072559 - 64094008 on Build GRCh38
- UniGene ID
- Hs.439548
- Species
- Homo sapiens
- Gene Symbol
- LOC439933
- Entrez Gene ID
- 439933
- Gene Name
- uncharacterized LOC439933
- Gene Aliases
- Chromosome Location
- Chr. 4: 36244116 - 36274215 on Build GRCh38
- UniGene ID
- Hs.591071
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_122080.1 |
|
2 |
276 |
|
GenBank mRNA |
BC030111.1 |
- |
2 |
282 |
|
- Gene Symbol
- SHC1
- Entrez Gene ID
- 6464
- Gene Name
- SHC adaptor protein 1
- Gene Aliases
- SHC, SHCA
- Chromosome Location
- Chr. 1: 154962298 - 154974492 on Build GRCh38
- UniGene ID
- Hs.433795
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB208849.1 |
- |
8 |
1747 |
|
View Full Product Details