Gene Transcripts

Gene Symbol
CARD17
Entrez Gene ID
440068
Gene Name
caspase recruitment domain family member 17
Gene Aliases
INCA
Chromosome Location
Chr. 11: 105092469 - 105101487 on Build GRCh38
UniGene ID
Hs.732627
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007232.1 NP_001007233.1 4 403
GenBank mRNA AY761064.1 - 4 403

View Full Product Details