Gene Transcripts
- Gene Symbol
- FCGR1B
- Entrez Gene ID
- 2210
- Gene Name
- Fc fragment of IgG receptor Ib
- Gene Aliases
- CD64b, IGFRB
- Chromosome Location
- Chr. 1: 121087345 - 121097161 on Build GRCh38
- UniGene ID
- Hs.534956
- Species
- Homo sapiens
- Gene Symbol
- FCGR1A
- Entrez Gene ID
- 2209
- Gene Name
- Fc fragment of IgG receptor Ia
- Gene Aliases
- CD64, CD64A, FCRI, IGFR1
- Chromosome Location
- Chr. 1: 149782677 - 149792518 on Build GRCh38
- UniGene ID
- Hs.77424
- Species
- Homo sapiens
- Gene Symbol
- FCGR1CP
- Entrez Gene ID
- 100132417
- Gene Name
- Fc fragment of IgG receptor Ic, pseudogene
- Gene Aliases
- CD64c, FCGR1C, FCRIC, IGFR1, IGFRC
- Chromosome Location
- Chr. 1: 143874743 - 143883733 on Build GRCh38
- UniGene ID
- Hs.635062
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027484.2 |
|
6 |
1077 |
|
GenBank mRNA |
CA309927.1 |
- |
1 |
272 |
|
CA748521.1 |
- |
1 |
265 |
|
View Full Product Details