Gene Transcripts

Gene Symbol
AK4
Entrez Gene ID
205
Gene Name
adenylate kinase 4
Gene Aliases
AK 4, AK3, AK3L1, AK3L2
Chromosome Location
Chr. 1: 65147342 - 65232145 on Build GRCh38
UniGene ID
Hs.10862
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005353.2 NP_001005353.1 4 754
NM_013410.3 NP_037542.1 3 637
NM_203464.2 NP_982289.1 4 643
XM_017000612.1 XP_016856101.1 3 389
XM_017000613.1 XP_016856102.1 3 575
GenBank mRNA AK025926.1 - 3 601
AK309057.1 - 3 345
AK313611.1 - 3 601
BC016180.1 - 3 537 4637988
BC040224.1 - 3 513 5188877
BC066944.1 - 4 754 4819382
BC136886.1 - 3 478 9020877
BC136887.1 - 1 478 9020878
BC146653.1 - 4 628 40147793
BC148270.1 - 3 418 8860213
BF205061.1 - 1 599
BU164554.1 - 4 600
CR456830.1 - 3 396
DR003868.1 - 4 676
X60673.1 - 3 579

View Full Product Details