Gene Transcripts
- Gene Symbol
- HMP19
- Entrez Gene ID
- 51617
- Gene Name
- HMP19 protein
- Gene Aliases
- Chromosome Location
- Chr. 5: 174045604 - 174109179 on Build GRCh38
- UniGene ID
- Hs.559412
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK098398.1 |
- |
|
630 |
|
View Full Product Details