Gene Transcripts

Gene Symbol
OR8G1
Entrez Gene ID
26494
Gene Name
olfactory receptor family 8 subfamily G member 1 (gene/pseudogene)
Gene Aliases
HSTPCR25, OR8G1P, TPCR25
Chromosome Location
Chr. 11: 124249676 - 124250611 on Build GRCh38
UniGene ID
Hs.448172
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002905.1 NP_001002905.1 1 739
NR_045681.1 1 739
GenBank mRNA X89672.1 - 1 365

View Full Product Details