Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001244071.1 | NP_001231000.1 | 3 | 1052 | |
NM_001244072.1 | NP_001231001.1 | 3 | 1486 | ||
NM_024711.5 | NP_078987.3 | 3 | 1276 | ||
GenBank mRNA | AJ580952.1 | - | 3 | 750 | |
AK026343.1 | - | 3 | 723 | ||
AK074044.1 | - | 2 | 2467 | ||
AK294989.1 | - | 3 | 1121 | ||
AK300445.1 | - | 3 | 992 | ||
AK314667.1 | - | 3 | 980 | ||
BC060760.1 | - | 3 | 937 | 30336063 | |
BC074744.2 | - | 3 | 826 | 30915204 | |
CR749355.1 | - | 3 | 1013 |