Gene Transcripts
- Gene Symbol
- OBSCN
- Entrez Gene ID
- 84033
- Gene Name
- obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
- Gene Aliases
- ARHGEF30, UNC89
- Chromosome Location
- Chr. 1: 228208013 - 228381431 on Build GRCh38
- UniGene ID
- Hs.650039
- Species
- Homo sapiens
- Gene Symbol
- C1orf145
- Entrez Gene ID
- 574407
- Gene Name
- chromosome 1 open reading frame 145
- Gene Aliases
- Chromosome Location
- Chr. 1: 228203506 - 228213664 on Build GRCh38
- UniGene ID
- Hs.650039
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_073154.1 |
|
2 |
462 |
|
GenBank mRNA |
AK056556.1 |
- |
2 |
462 |
|
View Full Product Details