Gene Transcripts
- Gene Symbol
- GGT2
- Entrez Gene ID
- 728441
- Gene Name
- gamma-glutamyltransferase 2
- Gene Aliases
- GGT, GGT 2
- Chromosome Location
- Chr. 22: 21207972 - 21282982 on Build GRCh38
- UniGene ID
- Hs.568255
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_005261890.4 |
XP_005261947.1
|
6, 7 |
899 |
|
XM_006724392.3 |
XP_006724455.1
|
10, 11 |
1607 |
|
XM_006724395.3 |
XP_006724458.1
|
7, 8 |
1059 |
|
XM_006724396.2 |
XP_006724459.1
|
3, 4 |
437 |
|
XM_011530621.2 |
XP_011528923.1
|
3, 4 |
424 |
|
XM_017029179.1 |
XP_016884668.1
|
11, 12 |
2288 |
|
XM_017029180.1 |
XP_016884669.1
|
10, 11 |
1746 |
|
XM_017029181.1 |
XP_016884670.1
|
10, 11 |
2184 |
|
XM_017029182.1 |
XP_016884671.1
|
11, 12 |
2288 |
|
XM_017029183.1 |
XP_016884672.1
|
6, 7 |
875 |
|
XM_017029184.1 |
XP_016884673.1
|
3, 4 |
447 |
|
XM_017029185.1 |
XP_016884674.1
|
12, 13 |
3150 |
|
XM_017029186.1 |
XP_016884675.1
|
15, 16 |
2751 |
|
GenBank mRNA |
M30474.1 |
- |
2, 3 |
233 |
|
- Gene Symbol
- GGT3P
- Entrez Gene ID
- 2679
- Gene Name
- gamma-glutamyltransferase 3 pseudogene
- Gene Aliases
- GGT3
- Chromosome Location
- Chr. 22: 18773689 - 18791961 on Build GRCh38
- UniGene ID
- Hs.595809
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003267.1 |
|
10, 11 |
1539 |
|
GenBank mRNA |
BC143607.1 |
- |
6, 7 |
1285 |
|
- Gene Symbol
- LOC102724197
- Entrez Gene ID
- 102724197
- Gene Name
- inactive gamma-glutamyltranspeptidase 2-like
- Gene Aliases
- GGT2
- Chromosome Location
- Chr. 22|NT_187386: 65299 - 86956 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_006724921.3 |
XP_006724984.1
|
10, 11 |
2050 |
|
XM_006724924.3 |
XP_006724987.1
|
9, 10 |
1892 |
|
XM_006724928.3 |
XP_006724991.1
|
5, 6 |
1088 |
|
XM_006724929.2 |
XP_006724992.1
|
3, 4 |
437 |
|
XM_011546256.2 |
XP_011544558.1
|
11, 12 |
2184 |
|
XM_017030133.1 |
XP_016885622.1
|
11, 12 |
1615 |
|
XM_017030134.1 |
XP_016885623.1
|
11, 12 |
1618 |
|
XM_017030135.1 |
XP_016885624.1
|
11, 12 |
2043 |
|
XM_017030136.1 |
XP_016885625.1
|
12, 13 |
2085 |
|
XM_017030137.1 |
XP_016885626.1
|
12, 13 |
2082 |
|
XM_017030138.1 |
XP_016885627.1
|
12, 13 |
1605 |
|
XM_017030139.1 |
XP_016885628.1
|
12, 13 |
1608 |
|
XM_017030140.1 |
XP_016885629.1
|
13, 14 |
2075 |
|
XM_017030141.1 |
XP_016885630.1
|
13, 14 |
2072 |
|
XM_017030142.1 |
XP_016885631.1
|
11, 12 |
1916 |
|
XM_017030143.1 |
XP_016885632.1
|
10, 11 |
2051 |
|
XM_017030144.1 |
XP_016885633.1
|
6, 7 |
979 |
|
XM_017030145.1 |
XP_016885634.1
|
7, 8 |
1612 |
|
- Gene Symbol
- LOC102724823
- Entrez Gene ID
- 102724823
- Gene Name
- inactive gamma-glutamyltranspeptidase 2
- Gene Aliases
- Chromosome Location
- Chr. 22|NT_187389: 6 - 18670 on Build GRCh38
- Species
- Homo sapiens
View Full Product Details