Gene Transcripts

Gene Symbol
MEG3
Entrez Gene ID
55384
Gene Name
maternally expressed 3 (non-protein coding)
Gene Aliases
FP504, GTL2, LINC00023, NCRNA00023, PRO0518, PRO2160, onco-lncRNA-83, prebp1
Chromosome Location
Chr. 14: 100826108 - 100861026 on Build GRCh38
UniGene ID
Hs.654863
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002766.2
3 333
NR_003530.2
3 333
NR_003531.3
3 309
NR_033358.1
3 333
NR_033359.1
3 333
NR_033360.1
3 333
NR_046464.1
3 333
NR_046465.2
3 333
NR_046466.1
3 333
NR_046467.1
3 333
NR_046469.1
3 333
NR_046470.2
3 333
NR_046471.1
3 333
NR_046472.1
3 333
NR_046473.1
3 333
GenBank mRNA AB032607.1 -
3 322
AF052114.1 -
1 75
AY314975.1 - LIT3321
MEG3
3 309
GQ183494.1 -
3 325
GQ183495.1 -
3 325
GQ183496.1 -
3 325
GQ183497.1 -
3 325
GQ183498.1 -
3 325
GQ183499.1 -
3 325
GQ183500.1 -
3 325
GQ183501.1 -
3 325
GQ183502.1 -
3 325
GQ183503.1 -
3 325

View Full Product Details