Gene Transcripts
- Gene Symbol
- TTTY8
- Entrez Gene ID
- 84673
- Gene Name
- testis-specific transcript, Y-linked 8 (non-protein coding)
- Gene Aliases
- LINC00130, NCRNA00130, TTY8
- Chromosome Location
- Chr. Y: 9691100 - 9693699 on Build GRCh38
- UniGene ID
- Hs.742605
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001533.1 |
|
2 |
169 |
|
- Gene Symbol
- TTTY8B
- Entrez Gene ID
- 100101118
- Gene Name
- testis-specific transcript, Y-linked 8B (non-protein coding)
- Gene Aliases
- NCRNA00217, TTTY8
- Chromosome Location
- Chr. Y: 6471031 - 6473630 on Build GRCh38
- UniGene ID
- Hs.461138
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003591.1 |
|
2 |
169 |
|
GenBank mRNA |
AA620621.1 |
- |
2 |
361 |
|
View Full Product Details