Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_040584.2 | 1 | 95 | ||
GenBank mRNA | AK303473.1 | - | 1 | 49 | |
AK307228.1 | - | 1 | 49 | ||
AK309007.1 | - | 1 | 50 | ||
BC002581.2 | - | 1 | 37 | ||
BG537494.1 | - | 1 | 94 | ||
BI753182.1 | - | 1 | 40 | ||
CB956482.1 | - | 1 | 74 |