Gene Transcripts
- Gene Symbol
- LOC100128651
- Entrez Gene ID
- 100128651
- Gene Name
- uncharacterized LOC100128651
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.536770
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK127243.1 |
- |
1 |
195 |
|
- Gene Symbol
- C4orf50
- Entrez Gene ID
- 389197
- Gene Name
- chromosome 4 open reading frame 50
- Gene Aliases
- Chromosome Location
- Chr. 4: 5897591 - 6018762 on Build GRCh38
- UniGene ID
- Hs.479065
- Species
- Homo sapiens
View Full Product Details