Gene Transcripts

Gene Symbol
MFAP3L
Entrez Gene ID
9848
Gene Name
microfibrillar associated protein 3 like
Gene Aliases
NYD-sp9
Chromosome Location
Chr. 4: 169986597 - 170033028 on Build GRCh38
UniGene ID
Hs.593942
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017008865.1 XP_016864354.1 1 1549
XM_017008869.1 XP_016864358.1 1 1549
XM_017008870.1 XP_016864359.1 1 1549

View Full Product Details