Gene Transcripts

Gene Symbol
PRKCQ
Entrez Gene ID
5588
Gene Name
protein kinase C theta
Gene Aliases
PRKCT, nPKC-theta
Chromosome Location
Chr. 10: 6393038 - 6580646 on Build GRCh38
UniGene ID
Hs.498570
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006717465.3 XP_006717528.1 1 663
XM_017016410.1 XP_016871899.1 1 663
XM_017016411.1 XP_016871900.1 1 663

Gene Symbol
LOC100130920
Entrez Gene ID
100130920
Gene Name
uncharacterized LOC100130920
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.683904
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK093639.1 - 1 663

View Full Product Details