Gene Transcripts

Gene Symbol
MTA3
Entrez Gene ID
57504
Gene Name
metastasis associated 1 family member 3
Gene Aliases
Chromosome Location
Chr. 2: 42494569 - 42795032 on Build GRCh38
UniGene ID
Hs.435413
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282755.1 NP_001269684.1 1 165
XM_005264459.2 XP_005264516.1 1 165
GenBank mRNA AK127245.1 - 1 165
DA239202.1 - 1 165

View Full Product Details