Gene Transcripts

Gene Symbol
SAYSD1
Entrez Gene ID
55776
Gene Name
SAYSVFN motif domain containing 1
Gene Aliases
C6orf64
Chromosome Location
Chr. 6: 39104063 - 39115186 on Build GRCh38
UniGene ID
Hs.58382
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304793.1 NP_001291722.1 2 1606
GenBank mRNA CR627244.1 - 2 1490

View Full Product Details