Gene Transcripts

Gene Symbol
KRTAP16-1
Entrez Gene ID
100505753
Gene Name
keratin associated protein 16-1
Gene Aliases
KAP16.1
Chromosome Location
Chr. 17: 41307700 - 41309253 on Build GRCh38
UniGene ID
Hs.721600
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146182.1 NP_001139654.1 1 988

View Full Product Details