Gene Transcripts

Gene Symbol
ANKRD31
Entrez Gene ID
256006
Gene Name
ankyrin repeat domain 31
Gene Aliases
Chromosome Location
Chr. 5: 75068297 - 75236897 on Build GRCh38
UniGene ID
Hs.482533
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164443.1 NP_001157915.1 21 5331
XM_011543298.2 XP_011541600.1 22 5524
XM_011543299.1 XP_011541601.1 22 5502
XM_011543300.2 XP_011541602.1 22 5524
XM_011543301.2 XP_011541603.1 21 5353
XM_011543302.2 XP_011541604.1 20 5060
XM_017009317.1 XP_016864806.1 22 5419
XM_017009318.1 XP_016864807.1 22 5524
XM_017009319.1 XP_016864808.1 22 5233
XM_017009320.1 XP_016864809.1 19 5045
XM_017009321.1 XP_016864810.1 15 4109

View Full Product Details