Gene Transcripts

Gene Symbol
PARP2
Entrez Gene ID
10038
Gene Name
poly(ADP-ribose) polymerase 2
Gene Aliases
ADPRT2, ADPRTL2, ADPRTL3, ARTD2, PARP-2, pADPRT-2
Chromosome Location
Chr. 14: 20343041 - 20357904 on Build GRCh38
UniGene ID
Hs.409412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042618.1 NP_001036083.1 11 1010
NM_005484.3 NP_005475.2 11 1049
XM_005267247.3 XP_005267304.1 11 1049
XM_017020912.1 XP_016876401.1 11 1583
GenBank mRNA AF085734.1 - 11 1046
AJ236876.2 - 10 960
AJ236912.1 - 11 984
AK001980.1 - 11 1026
AK290079.1 - 11 989
AK300971.1 - 9 767

View Full Product Details