Gene Transcripts

Gene Symbol
AP1M2
Entrez Gene ID
10053
Gene Name
adaptor related protein complex 1 mu 2 subunit
Gene Aliases
AP1-mu2, HSMU1B, MU-1B, MU1B, mu2
Chromosome Location
Chr. 19: 10572671 - 10587315 on Build GRCh38
UniGene ID
Hs.18894
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300887.1 NP_001287816.1 1 105
NM_005498.4 NP_005489.2 1 105
GenBank mRNA AF020797.2 - 1 68
AK023863.1 - 1 106
AK222946.1 - 1 71
AK315689.1 - 1 96
BC003387.2 - 1 43 3446133
BC003612.2 - 1 42 2988458
BC005021.1 - 1 105 3139433

View Full Product Details