Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001144663.1 | NP_001138135.1 | 6 | 606 | |
NM_004063.3 | NP_004054.3 | 6 | 605 | ||
XM_011516790.1 | XP_011515092.1 | 6 | 604 | ||
GenBank mRNA | AK312873.1 | - | 6 | 606 | |
BC112013.1 | - | 6 | 578 | 8327481 | |
BC113464.1 | - | 6 | 566 | 8322516 | |
U07969.1 | - | 6 | 567 | ||
X83228.1 | - | 6 | 572 |