Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007225.2 | NP_009156.2 | 2 | 650 | |
GenBank mRNA | AB032985.1 | - | 1 | 274 | |
AF043468.1 | - | 1 | 129 | ||
AK308928.1 | - | 2 | 464 | ||
AL834282.1 | - | 2 | 728 | ||
AY358526.1 | - | 2 | 420 | ||
BC022541.1 | - | 2 | 469 | 4794968 |