Gene Transcripts
- Gene Symbol
- PALM2
- Entrez Gene ID
- 114299
- Gene Name
- paralemmin 2
- Gene Aliases
- AKAP2
- Chromosome Location
- Chr. 9: 109640788 - 109951476 on Build GRCh38
- UniGene ID
- Hs.591908
- Species
- Homo sapiens
- Gene Symbol
- PALM2-AKAP2
- Entrez Gene ID
- 445815
- Gene Name
- PALM2-AKAP2 readthrough
- Gene Aliases
- AKAP2
- Chromosome Location
- Chr. 9: 109780297 - 110172512 on Build GRCh38
- UniGene ID
- Hs.591908
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AJ312217.1 |
- |
7 |
866 |
|
View Full Product Details