Gene Transcripts
- Gene Symbol
- PKD1L2
- Entrez Gene ID
- 114780
- Gene Name
- polycystin 1 like 2 (gene/pseudogene)
- Gene Aliases
- PC1L2
- Chromosome Location
- Chr. 16: 81100879 - 81220370 on Build GRCh38
- UniGene ID
- Hs.413525
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001076780.1 |
NP_001070248.1
|
16, 17 |
2781 |
|
NM_001278423.1 |
NP_001265352.1
|
5, 6 |
875 |
|
NM_001278425.1 |
NP_001265354.1
|
5, 6 |
875 |
|
NM_052892.3 |
NP_443124.3
|
16, 17 |
2781 |
|
NR_126532.1 |
|
16, 17 |
2781 |
|
GenBank mRNA |
AB067466.1 |
- |
8, 9 |
1126 |
|
AK098052.1 |
- |
10, 11 |
1363 |
|
AY164483.1 |
- |
16, 17 |
2781 |
|
AY371495.1 |
- |
5, 6 |
837 |
|
BC004562.2 |
- |
5, 6 |
842 |
|
BC014157.2 |
- |
5, 6 |
842 |
|
View Full Product Details