Gene Transcripts
- Gene Symbol
- CKB
- Entrez Gene ID
- 1152
- Gene Name
- creatine kinase B
- Gene Aliases
- B-CK, BCK, CKBB, HEL-211, HEL-S-29
- Chromosome Location
- Chr. 14: 103519658 - 103522859 on Build GRCh38
- UniGene ID
- Hs.173724
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001823.4 |
NP_001814.2
|
6, 7 |
891 |
|
XM_017020951.1 |
XP_016876440.1
|
5, 6 |
937 |
|
GenBank mRNA |
AK290101.1 |
- |
6, 7 |
862 |
|
AK312282.1 |
- |
6, 7 |
863 |
|
BC001190.1 |
- |
6, 7 |
850 |
3354679 |
BC004914.2 |
- |
6, 7 |
842 |
3537262 |
BC008323.1 |
- |
6, 7 |
848 |
3358085 |
BC010002.2 |
- |
6, 7 |
857 |
4135106 |
BC019259.1 |
- |
6, 7 |
846 |
3503057 |
BC019281.1 |
- |
6, 7 |
856 |
2823530 |
BM684665.1 |
- |
2, 3 |
581 |
|
CR542268.1 |
- |
5, 6 |
785 |
|
EU794629.1 |
- |
6, 7 |
846 |
|
FJ224337.1 |
- |
5, 6 |
785 |
|
KU177977.1 |
- |
5, 6 |
785 |
|
KU177978.1 |
- |
4, 5 |
857 |
|
L47647.1 |
- |
6, 7 |
828 |
|
M16364.1 |
- |
5, 6 |
793 |
|
M16451.1 |
- |
5, 6 |
808 |
|
View Full Product Details