Gene Transcripts

Gene Symbol
DNAJC24
Entrez Gene ID
120526
Gene Name
DnaJ heat shock protein family (Hsp40) member C24
Gene Aliases
DPH4, JJJ3, ZCSL3
Chromosome Location
Chr. 11: 31369829 - 31432835 on Build GRCh38
UniGene ID
Hs.187269
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181706.4 NP_859057.4 3, 4 367
XM_017017216.1 XP_016872705.1 3, 4 352
GenBank mRNA AK289665.1 - 3, 4 355
AL833128.1 - 3, 4 357
BC063804.1 - 3, 4 310 6474523
CD685768.1 - 3, 4 385

View Full Product Details