Gene Transcripts

Gene Symbol
IKBIP
Entrez Gene ID
121457
Gene Name
IKBKB interacting protein
Gene Aliases
IKIP
Chromosome Location
Chr. 12: 98613404 - 98645051 on Build GRCh38
UniGene ID
Hs.252543
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153687.3 NP_710154.1 3 1110
GenBank mRNA AJ539425.1 - 3 870
AK055613.1 - 3 825
BC058933.1 - 3 818 5541278

View Full Product Details