Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152377.2 | NP_689590.1 | 5 | 799 | |
XM_017000307.1 | XP_016855796.1 | 4 | 609 | ||
XM_017000308.1 | XP_016855797.1 | 5 | 790 | ||
GenBank mRNA | AK124828.1 | - | 5 | 784 | |
BC027878.1 | - | 5 | 783 | 5207590 |