Gene Transcripts
- Gene Symbol
- NKAIN4
- Entrez Gene ID
- 128414
- Gene Name
- Na+/K+ transporting ATPase interacting 4
- Gene Aliases
- C20orf58, FAM77A, bA261N11.2
- Chromosome Location
- Chr. 20: 63240784 - 63254904 on Build GRCh38
- UniGene ID
- Hs.46627
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_152864.3 |
NP_690603.3
|
2, 3 |
272 |
|
XM_005260192.1 |
XP_005260249.1
|
2, 3 |
234 |
|
XM_005260194.3 |
XP_005260251.1
|
2, 3 |
693 |
|
XM_011528527.1 |
XP_011526829.1
|
2, 3 |
234 |
|
XM_011528528.2 |
XP_011526830.1
|
2, 3 |
696 |
|
XM_011528529.2 |
XP_011526831.1
|
2, 3 |
682 |
|
XM_017027636.1 |
XP_016883125.1
|
2, 3 |
204 |
|
XM_017027637.1 |
XP_016883126.1
|
2, 3 |
686 |
|
XM_017027638.1 |
XP_016883127.1
|
2, 3 |
692 |
|
XM_017027639.1 |
XP_016883128.1
|
2, 3 |
678 |
|
GenBank mRNA |
BC041812.1 |
- |
2, 3 |
270 |
|
BG708360.1 |
- |
2, 3 |
276 |
|
View Full Product Details